Assay Details
Mature miRNA Sequence: | CUCCGGGACGGCUGGGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4497 | MIMAT0019032 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4497 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0016859 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.12: 109833348 - 109833436 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | CUCCGGGACGGCUGGGC | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | CUCCGGGACGGCUGGGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4497 | MIMAT0019032 |
Stem-loop ID | hsa-mir-4497 |
Stem-loop Accession # | MI0016859 |
Stem-loop Sequence | ACCUCCGGGACGGCUGGGCGCCGGCGGCCGGGAGAUCCGCGCUUCCUGAAUCCCGGCCGGCCCGCCCGGCGCCCGUCCGCCCGCGGGUC |
Chromosome Location | Chr. 12 - 109833348 - 109833436 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019032
miRBase ID: hsa-miR-4497 Mature miRNA Sequence: CUCCGGGACGGCUGGGC Chromosome Location: Chr. 12 - 109833348 - 109833436 [+] on Build GRCh38 |