Assay Details
Gene Family ID: | MIPF0001935, mir-3149 |
Mature miRNA Sequence: | UUUGUAUGGAUAUGUGUGUGUAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3149 | MIMAT0015022 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3149 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0014176 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.8: 76966768 - 76966850 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UUUGUAUGGAUAUGUGUGUGUAU | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0001935, mir-3149 |
Mature miRNA Sequence: | UUUGUAUGGAUAUGUGUGUGUAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3149 | MIMAT0015022 |
Stem-loop ID | hsa-mir-3149 |
Stem-loop Accession # | MI0014176 |
Stem-loop Sequence | AUACAUACAUGUACACACACAUGUCAUCCACACACAUACAUAUAUAUAUGUUUGUAUGGAUAUGUGUGUGUAUGUGUGUGUAU |
Chromosome Location | Chr. 8 - 76966768 - 76966850 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0015022
miRBase ID: hsa-miR-3149 Mature miRNA Sequence: UUUGUAUGGAUAUGUGUGUGUAU Chromosome Location: Chr. 8 - 76966768 - 76966850 [-] on Build GRCh38 |