Assay Details
Gene Family ID: | MIPF0001615, mir-3140 |
Mature miRNA Sequence: | AGCUUUUGGGAAUUCAGGUAGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3140-3p | MIMAT0015008 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3140-3p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0014163 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.4: 152489327 - 152489416 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | AGCUUUUGGGAAUUCAGGUAGU | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0001615, mir-3140 |
Mature miRNA Sequence: | AGCUUUUGGGAAUUCAGGUAGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3140-3p | MIMAT0015008 |
Stem-loop ID | hsa-mir-3140 |
Stem-loop Accession # | MI0014163 |
Stem-loop Sequence | CCUCUUGAGGUACCUGAAUUACCAAAAGCUUUAUGUAUUCUGAAGUUAUUGAAAAUAAGAGCUUUUGGGAAUUCAGGUAGUUCAGGAGUG |
Chromosome Location | Chr. 4 - 152489327 - 152489416 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0015008
miRBase ID: hsa-miR-3140-3p Mature miRNA Sequence: AGCUUUUGGGAAUUCAGGUAGU Chromosome Location: Chr. 4 - 152489327 - 152489416 [-] on Build GRCh38 |