Assay Details
Gene Family ID: | MIPF0000679, mir-641 |
Mature miRNA Sequence: | AAAGACAUAGGAUAGAGUCACCUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-641 | MIMAT0003311 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-641 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0003656 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.19: 40282543 - 40282641 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | AAAGACAUAGGAUAGAGUCACCUC | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000679, mir-641 |
Mature miRNA Sequence: | AAAGACAUAGGAUAGAGUCACCUC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-641 | MIMAT0003311 |
Stem-loop ID | hsa-mir-641 |
Stem-loop Accession # | MI0003656 |
Stem-loop Sequence | UGGGUGAAAGGAAGGAAAGACAUAGGAUAGAGUCACCUCUGUCCUCUGUCCUCUACCUAUAGAGGUGACUGUCCUAUGUCUUUCCUUCCUCUUACCCCU |
Chromosome Location | Chr. 19 - 40282543 - 40282641 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0003311
miRBase ID: hsa-miR-641 Mature miRNA Sequence: AAAGACAUAGGAUAGAGUCACCUC Chromosome Location: Chr. 19 - 40282543 - 40282641 [-] on Build GRCh38 |