Assay Details
Gene Family ID: | MIPF0000503, mir-593 |
Mature miRNA Sequence: | AGGCACCAGCCAGGCAUUGCUCAGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-593-5p | MIMAT0003261 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-593-5p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0003605 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.7: 128081861 - 128081960 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AGGCACCAGCCAGGCAUUGCUCAGC | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000503, mir-593 |
Mature miRNA Sequence: | AGGCACCAGCCAGGCAUUGCUCAGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-593-5p | MIMAT0003261 |
Stem-loop ID | hsa-mir-593 |
Stem-loop Accession # | MI0003605 |
Stem-loop Sequence | CCCCCAGAAUCUGUCAGGCACCAGCCAGGCAUUGCUCAGCCCGUUUCCCUCUGGGGGAGCAAGGAGUGGUGCUGGGUUUGUCUCUGCUGGGGUUUCUCCU |
Chromosome Location | Chr. 7 - 128081861 - 128081960 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0003261
miRBase ID: hsa-miR-593-5p Mature miRNA Sequence: AGGCACCAGCCAGGCAUUGCUCAGC Chromosome Location: Chr. 7 - 128081861 - 128081960 [+] on Build GRCh38 |