Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | UGGCAGUUACUUUUGCACCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548au-3p | MIMAT0022292 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-548au-3p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0019145 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.9: 93594841 - 93594894 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UGGCAGUUACUUUUGCACCAG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | UGGCAGUUACUUUUGCACCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548au-3p | MIMAT0022292 |
Stem-loop ID | hsa-mir-548au |
Stem-loop Accession # | MI0019145 |
Stem-loop Sequence | AAAAGUAAUUGCGGUUUUUGCUAUUGGUUUUAAUGGCAGUUACUUUUGCACCAG |
Chromosome Location | Chr. 9 - 93594841 - 93594894 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0022292
miRBase ID: hsa-miR-548au-3p Mature miRNA Sequence: UGGCAGUUACUUUUGCACCAG Chromosome Location: Chr. 9 - 93594841 - 93594894 [+] on Build GRCh38 |