Assay Details
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | UUCUCAAGAGGGAGGCAAUCAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-514b-5p | MIMAT0015087 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-514b-5p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0014251 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.X: 147250151 - 147250230 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UUCUCAAGAGGGAGGCAAUCAU | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | UUCUCAAGAGGGAGGCAAUCAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-514b-5p | MIMAT0015087 |
Stem-loop ID | hsa-mir-514b |
Stem-loop Accession # | MI0014251 |
Stem-loop Sequence | CAUGUGGUACUCUUCUCAAGAGGGAGGCAAUCAUGUGUAAUUAGAUAUGAUUGACACCUCUGUGAGUGGAGUAACACAUG |
Chromosome Location | Chr. X - 147250151 - 147250230 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0015087
miRBase ID: hsa-miR-514b-5p Mature miRNA Sequence: UUCUCAAGAGGGAGGCAAUCAU Chromosome Location: Chr. X - 147250151 - 147250230 [-] on Build GRCh38 |