Assay Details
Mature miRNA Sequence: | UAGGAUUACAAGUGUCGGCCAC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3159 | MIMAT0015033 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3159 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0014188 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.11: 18387787 - 18387860 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UAGGAUUACAAGUGUCGGCCAC | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UAGGAUUACAAGUGUCGGCCAC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3159 | MIMAT0015033 |
Stem-loop ID | hsa-mir-3159 |
Stem-loop Accession # | MI0014188 |
Stem-loop Sequence | CCAAAGUCCUAGGAUUACAAGUGUCGGCCACGGGCUGGGCACAGUGGCUCACGCCUGUAAUCCCAGCAUUUUGG |
Chromosome Location | Chr. 11 - 18387787 - 18387860 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0015033
miRBase ID: hsa-miR-3159 Mature miRNA Sequence: UAGGAUUACAAGUGUCGGCCAC Chromosome Location: Chr. 11 - 18387787 - 18387860 [+] on Build GRCh38 |