Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CAAAAACCACAGUUUCUUUUGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548d-3p | MIMAT0003323 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-548d | ||
---|---|---|---|
miRBase Accession Number: | MI0003668 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.8: 123348034 - 123348130 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | CAAAAACCACAGUUUCUUUUGC | ||
Species: | Human | ||
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CAAAAACCACAGUUUCUUUUGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548d-3p | MIMAT0003323 |
Stem-loop ID | hsa-mir-548d-1 |
Stem-loop Accession # | MI0003668 |
Stem-loop Sequence | AAACAAGUUAUAUUAGGUUGGUGCAAAAGUAAUUGUGGUUUUUGCCUGUAAAAGUAAUGGCAAAAACCACAGUUUCUUUUGCACCAGACUAAUAAAG |
Chromosome Location | Chr. 8 - 123348034 - 123348130 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0003323
miRBase ID: hsa-miR-548d-3p Mature miRNA Sequence: CAAAAACCACAGUUUCUUUUGC Chromosome Location: Chr. 8 - 123348034 - 123348130 [-] on Build GRCh38 |
Stem-loop ID | hsa-mir-548d-2 |
Stem-loop Accession # | MI0003671 |
Stem-loop Sequence | GAGAGGGAAGAUUUAGGUUGGUGCAAAAGUAAUUGUGGUUUUUGCCAUUGAAAGUAAUGGCAAAAACCACAGUUUCUUUUGCACCAACCUAAUAAAA |
Chromosome Location | Chr. 17 - 67471489 - 67471585 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0003323
miRBase ID: hsa-miR-548d-3p Mature miRNA Sequence: CAAAAACCACAGUUUCUUUUGC Chromosome Location: Chr. 8 - 123348034 - 123348130 [-] on Build GRCh38 |