Assay Details
Mature miRNA Sequence: | UCCUGUCUUUCCUUGUUGGAGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5699-3p | MIMAT0022492 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-5699-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0019306 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.10: 641689 - 641778 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UCCUGUCUUUCCUUGUUGGAGC | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | UCCUGUCUUUCCUUGUUGGAGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-5699-3p | MIMAT0022492 |
Stem-loop ID | hsa-mir-5699 |
Stem-loop Accession # | MI0019306 |
Stem-loop Sequence | CUGUACCCCUGCCCCAACAAGGAAGGACAAGAGGUGUGAGCCACACACACGCCUGGCCUCCUGUCUUUCCUUGUUGGAGCAGGGAUGUAG |
Chromosome Location | Chr. 10 - 641689 - 641778 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0022492
miRBase ID: hsa-miR-5699-3p Mature miRNA Sequence: UCCUGUCUUUCCUUGUUGGAGC Chromosome Location: Chr. 10 - 641689 - 641778 [-] on Build GRCh38 |