Assay Details
Mature miRNA Sequence: | UCUGCCAUCCUCCCUCCCCUAC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4769-3p | MIMAT0019923 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4769-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0017410 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.X: 47587429 - 47587505 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UCUGCCAUCCUCCCUCCCCUAC | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | UCUGCCAUCCUCCCUCCCCUAC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4769-3p | MIMAT0019923 |
Stem-loop ID | hsa-mir-4769 |
Stem-loop Accession # | MI0017410 |
Stem-loop Sequence | GAGGAGAGGUGGGAUGGAGAGAAGGUAUGAGCUAAAAAUCCCCAAGCUCUGCCAUCCUCCCUCCCCUACUUCUCCCC |
Chromosome Location | Chr. X - 47587429 - 47587505 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019923
miRBase ID: hsa-miR-4769-3p Mature miRNA Sequence: UCUGCCAUCCUCCCUCCCCUAC Chromosome Location: Chr. X - 47587429 - 47587505 [+] on Build GRCh38 |