Assay Details
Mature miRNA Sequence: | UGCUUAAGUUGUACCAAGUAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4720-3p | MIMAT0019834 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4720-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0017355 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.16: 81385018 - 81385093 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UGCUUAAGUUGUACCAAGUAU | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | UGCUUAAGUUGUACCAAGUAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4720-3p | MIMAT0019834 |
Stem-loop ID | hsa-mir-4720 |
Stem-loop Accession # | MI0017355 |
Stem-loop Sequence | AAGCCUGGCAUAUUUGGUAUAACUUAAGCACCAGGUAAAAUCUGGUGCUUAAGUUGUACCAAGUAUAGCCAAGUUU |
Chromosome Location | Chr. 16 - 81385018 - 81385093 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019834
miRBase ID: hsa-miR-4720-3p Mature miRNA Sequence: UGCUUAAGUUGUACCAAGUAU Chromosome Location: Chr. 16 - 81385018 - 81385093 [+] on Build GRCh38 |