Assay Details
Mature miRNA Sequence: | UGUCCUCUAGGGCCUGCAGUCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3909 | MIMAT0018183 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3909 | ||
---|---|---|---|
miRBase Accession Number: | MI0016413 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.22: 35335640 - 35335758 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UGUCCUCUAGGGCCUGCAGUCU | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | UGUCCUCUAGGGCCUGCAGUCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3909 | MIMAT0018183 |
Stem-loop ID | hsa-mir-3909 |
Stem-loop Accession # | MI0016413 |
Stem-loop Sequence | GGUAUGCUGUUGCGCUGUCCUUCCUCUGGGGAGCAGGCUCCGGGGGACAGGGAAAAGCACACAAGGAACUUGUCCUCUAGGGCCUGCAGUCUCAUGGGAGAGUGACAUGCACCAGGACC |
Chromosome Location | Chr. 22 - 35335640 - 35335758 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0018183
miRBase ID: hsa-miR-3909 Mature miRNA Sequence: UGUCCUCUAGGGCCUGCAGUCU Chromosome Location: Chr. 22 - 35335640 - 35335758 [+] on Build GRCh38 |