Assay Details
Gene Family ID: | MIPF0000316, mir-467 |
Mature miRNA Sequence: | AUACACAUACACGCAACACACAU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-466 | MIMAT0015002 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-466 | ||
---|---|---|---|
miRBase Accession Number: | MI0014157 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.3: 31161704 - 31161787 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | AUACACAUACACGCAACACACAU | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000316, mir-467 |
Mature miRNA Sequence: | AUACACAUACACGCAACACACAU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-466 | MIMAT0015002 |
Stem-loop ID | hsa-mir-466 |
Stem-loop Accession # | MI0014157 |
Stem-loop Sequence | GUGUGUGUAUAUGUGUGUUGCAUGUGUGUAUAUGUGUGUAUAUAUGUACACAUACACAUACACGCAACACACAUAUAUACAUGC |
Chromosome Location | Chr. 3 - 31161704 - 31161787 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0015002
miRBase ID: hsa-miR-466 Mature miRNA Sequence: AUACACAUACACGCAACACACAU Chromosome Location: Chr. 3 - 31161704 - 31161787 [-] on Build GRCh38 |