Assay Details
Gene Family ID: | MIPF0000972, mir-608 |
Mature miRNA Sequence: | AGGGGUGGUGUUGGGACAGCUCCGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-608 | MIMAT0003276 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-608 | ||
---|---|---|---|
miRBase Accession Number: | MI0003621 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.10: 100974985 - 100975084 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AGGGGUGGUGUUGGGACAGCUCCGU | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000972, mir-608 |
Mature miRNA Sequence: | AGGGGUGGUGUUGGGACAGCUCCGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-608 | MIMAT0003276 |
Stem-loop ID | hsa-mir-608 |
Stem-loop Accession # | MI0003621 |
Stem-loop Sequence | GGGCCAAGGUGGGCCAGGGGUGGUGUUGGGACAGCUCCGUUUAAAAAGGCAUCUCCAAGAGCUUCCAUCAAAGGCUGCCUCUUGGUGCAGCACAGGUAGA |
Chromosome Location | Chr. 10 - 100974985 - 100975084 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0003276
miRBase ID: hsa-miR-608 Mature miRNA Sequence: AGGGGUGGUGUUGGGACAGCUCCGU Chromosome Location: Chr. 10 - 100974985 - 100975084 [+] on Build GRCh38 |