Assay Details
Mature miRNA Sequence: | UGGCUUCUCUUGCACACCCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6815-3p | MIMAT0027531 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6815-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0022660 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.21: 45478266 - 45478326 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UGGCUUCUCUUGCACACCCAG | ||
Species: | Human | ||
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UGGCUUCUCUUGCACACCCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6815-3p | MIMAT0027531 |
Stem-loop ID | hsa-mir-6815 |
Stem-loop Accession # | MI0022660 |
Stem-loop Sequence | CACUGUAGGUGGCGCCGGAGGAGUCAUUUCCCAUCACUAAUGGCUUCUCUUGCACACCCAG |
Chromosome Location | Chr. 21 - 45478266 - 45478326 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027531
miRBase ID: hsa-miR-6815-3p Mature miRNA Sequence: UGGCUUCUCUUGCACACCCAG Chromosome Location: Chr. 21 - 45478266 - 45478326 [+] on Build GRCh38 |