Assay Details
Mature miRNA Sequence: | GGGAAGAGCUGUACGGCCUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6077 | MIMAT0023702 |
Stem-loop Details
Human
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6077 | ||
---|---|---|---|
miRBase Accession Number: | MI0020354 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.1: 148388282 - 148388363 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | GGGAAGAGCUGUACGGCCUUC | ||
Species: | Human | ||
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | GGGAAGAGCUGUACGGCCUUC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6077 | MIMAT0023702 |
Stem-loop ID | hsa-mir-6077 |
Stem-loop Accession # | MI0020354 |
Stem-loop Sequence | CCUGCGUAGAAAGGGAAGAGCUGUACGGCCUUCGCGCUAGUUUGUUUGUGUUAAGGCUGACGCUCCCUAUUUCUCCUCAGUG |
Chromosome Location | Chr. 1 - 148388282 - 148388363 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0023702
miRBase ID: hsa-miR-6077 Mature miRNA Sequence: GGGAAGAGCUGUACGGCCUUC Chromosome Location: Chr. 1 - 148388282 - 148388363 [+] on Build GRCh38 |