Assay Details
Mature miRNA Sequence: | UGCUGUAUUGUCAGGUAGUGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4999-5p | MIMAT0021017 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4999-5p | ||
---|---|---|---|
miRBase Accession Number: | MI0017865 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.19: 8389290 - 8389380 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UGCUGUAUUGUCAGGUAGUGA | ||
Species: | Human | ||
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | UGCUGUAUUGUCAGGUAGUGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4999-5p | MIMAT0021017 |
Stem-loop ID | hsa-mir-4999 |
Stem-loop Accession # | MI0017865 |
Stem-loop Sequence | AUAGAAAAUAAAACACAUACUGCUGUAUUGUCAGGUAGUGAUAGGAUUUAUCACUACCUGACAAUACAGUAUGUGUUUGUUUUAUAUAUUU |
Chromosome Location | Chr. 19 - 8389290 - 8389380 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0021017
miRBase ID: hsa-miR-4999-5p Mature miRNA Sequence: UGCUGUAUUGUCAGGUAGUGA Chromosome Location: Chr. 19 - 8389290 - 8389380 [-] on Build GRCh38 |