Assay Details
Mature miRNA Sequence: | CUGCAGCCACUUGGGGAACUGGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-7156-3p | MIMAT0028223 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-7156-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0023616 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.1: 77060143 - 77060202 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | CUGCAGCCACUUGGGGAACUGGU | ||
Species: | Human | ||
Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | CUGCAGCCACUUGGGGAACUGGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-7156-3p | MIMAT0028223 |
Stem-loop ID | hsa-mir-7156 |
Stem-loop Accession # | MI0023616 |
Stem-loop Sequence | UUGUUCUCAAACUGGCUGUCAGAGUGUGCAUGGCAGGCUGCAGCCACUUGGGGAACUGGU |
Chromosome Location | Chr. 1 - 77060143 - 77060202 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0028223
miRBase ID: hsa-miR-7156-3p Mature miRNA Sequence: CUGCAGCCACUUGGGGAACUGGU Chromosome Location: Chr. 1 - 77060143 - 77060202 [+] on Build GRCh38 |