Assay Details
Mature miRNA Sequence: | UGCCCAUGCCAUACUUUUGCCUCA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4641 | MIMAT0019701 |
Stem-loop Details
Human
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4641 | ||
---|---|---|---|
miRBase Accession Number: | MI0017268 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.6: 41598723 - 41598788 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UGCCCAUGCCAUACUUUUGCCUCA | ||
Species: | Human | ||
Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UGCCCAUGCCAUACUUUUGCCUCA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4641 | MIMAT0019701 |
Stem-loop ID | hsa-mir-4641 |
Stem-loop Accession # | MI0017268 |
Stem-loop Sequence | GGGGGGCAGGGGGCAGAGGGCAUCAGAGGACAGCCGCCUGGUGCCCAUGCCAUACUUUUGCCUCAG |
Chromosome Location | Chr. 6 - 41598723 - 41598788 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019701
miRBase ID: hsa-miR-4641 Mature miRNA Sequence: UGCCCAUGCCAUACUUUUGCCUCA Chromosome Location: Chr. 6 - 41598723 - 41598788 [+] on Build GRCh38 |