Assay Details
Mature miRNA Sequence: | UCCGUACAAACUCUGCUGUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3678-5p | MIMAT0018102 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3678-5p | ||
---|---|---|---|
miRBase Accession Number: | MI0016079 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.17: 75406069 - 75406162 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UCCGUACAAACUCUGCUGUG | ||
Species: | Human | ||
Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UCCGUACAAACUCUGCUGUG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3678-5p | MIMAT0018102 |
Stem-loop ID | hsa-mir-3678 |
Stem-loop Accession # | MI0016079 |
Stem-loop Sequence | GAAUCCGGUCCGUACAAACUCUGCUGUGUUGAAUGAUUGGUGAGUUUGUUUGCUCAUUGAUUGAAUCACUGCAGAGUUUGUACGGACCGGAUUC |
Chromosome Location | Chr. 17 - 75406069 - 75406162 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0018102
miRBase ID: hsa-miR-3678-5p Mature miRNA Sequence: UCCGUACAAACUCUGCUGUG Chromosome Location: Chr. 17 - 75406069 - 75406162 [+] on Build GRCh38 |