Assay Details
Amplicon Length: | 108 |
Chromosome Location: | Chr. 6 - 44253959 - 44253959 [-] on 44253959 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4647 | MIMAT0019709 |
Stem-loop Details
Human