Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. 8 - 144393851 - 144393851 [+] on 144393851 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-939-5p | MIMAT0004982 |
Human | hsa-miR-939-3p | MIMAT0022939 |
Stem-loop Details
Human