Assay Details
Mature miRNA Sequence: | UAGGGGGCGGCUUGUGGAGUGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6805-5p | MIMAT0027510 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6805-5p | ||
---|---|---|---|
miRBase Accession Number: | MI0022650 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.19: 55388181 - 55388242 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UAGGGGGCGGCUUGUGGAGUGU | ||
Species: | Human | ||
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | UAGGGGGCGGCUUGUGGAGUGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6805-5p | MIMAT0027510 |
Stem-loop ID | hsa-mir-6805 |
Stem-loop Accession # | MI0022650 |
Stem-loop Sequence | UGGCCUAGGGGGCGGCUUGUGGAGUGUAUGGGCUGAGCCUUGCUCUGCUCCCCCGCCCCCAG |
Chromosome Location | Chr. 19 - 55388181 - 55388242 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027510
miRBase ID: hsa-miR-6805-5p Mature miRNA Sequence: UAGGGGGCGGCUUGUGGAGUGU Chromosome Location: Chr. 19 - 55388181 - 55388242 [+] on Build GRCh38 |