Assay Details
Mature miRNA Sequence: | AUACCUCAUCUAGAAUGCUGUA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4777-3p | MIMAT0019935 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4777-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0017421 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.2: 231362708 - 231362793 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AUACCUCAUCUAGAAUGCUGUA | ||
Species: | Human | ||
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | AUACCUCAUCUAGAAUGCUGUA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4777-3p | MIMAT0019935 |
Stem-loop ID | hsa-mir-4777 |
Stem-loop Accession # | MI0017421 |
Stem-loop Sequence | UAGAAUAUUUCGGCAUUCUAGAUGAGAGAUAUAUAUAUACCUCAUAUGUAUAUGGUAUACCUCAUCUAGAAUGCUGUAAUAUUCUA |
Chromosome Location | Chr. 2 - 231362708 - 231362793 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019935
miRBase ID: hsa-miR-4777-3p Mature miRNA Sequence: AUACCUCAUCUAGAAUGCUGUA Chromosome Location: Chr. 2 - 231362708 - 231362793 [+] on Build GRCh38 |