Assay Details
Mature miRNA Sequence: | AGAGCUGGCUGAAGGGCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4487 | MIMAT0019021 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4487 | ||
---|---|---|---|
miRBase Accession Number: | MI0016848 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.11: 47400970 - 47401042 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AGAGCUGGCUGAAGGGCAG | ||
Species: | Human | ||
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | AGAGCUGGCUGAAGGGCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4487 | MIMAT0019021 |
Stem-loop ID | hsa-mir-4487 |
Stem-loop Accession # | MI0016848 |
Stem-loop Sequence | ACUGUCCUUCAGCCAGAGCUGGCUGAAGGGCAGAAGGGAACUGUCCUUCAGCCAGAGCUGGCUGAAGGGCAGA |
Chromosome Location | Chr. 11 - 47400970 - 47401042 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019021
miRBase ID: hsa-miR-4487 Mature miRNA Sequence: AGAGCUGGCUGAAGGGCAG Chromosome Location: Chr. 11 - 47400970 - 47401042 [+] on Build GRCh38 |