Assay Details
Mature miRNA Sequence: | UUCUGAGCUGAGGACAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4303 | MIMAT0016856 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4303 | ||
---|---|---|---|
Stem-loop Accession Number: | MI0015834 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.12: 97995383 - 97995448 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UUCUGAGCUGAGGACAG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | UUCUGAGCUGAGGACAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4303 | MIMAT0016856 |
Stem-loop ID | hsa-mir-4303 |
Stem-loop Accession # | MI0015834 |
Stem-loop Sequence | AGAAAAUAGCUUCUGAGCUGAGGACAGCUUGCUCUGCUUUUCUUUAGCUUAGGAGCUAACCAUGGU |
Chromosome Location | Chr. 12 - 97995383 - 97995448 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0016856
miRBase ID: hsa-miR-4303 Mature miRNA Sequence: UUCUGAGCUGAGGACAG Chromosome Location: Chr. 12 - 97995383 - 97995448 [-] on Build GRCh38 |