Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUAUUAUACAGUCAACCUCU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-656-3p | MIMAT0003332 |
Bovine | bta-miR-656 | MIMAT0009361 |
Dasypus novemcinctus | dno-miR-656-3p | MIMAT0047993 |
Goat | chi-miR-656 | MIMAT0036283 |
Guinea pig | cpo-miR-656-3p | MIMAT0047364 |
Horse | eca-miR-656 | MIMAT0013172 |
View More Rows | ||
Pan troglodytes | ptr-miR-656 | MIMAT0008305 |
Pongo pygmaeus | ppy-miR-656 | MIMAT0016090 |
Pteropus alecto | pal-miR-656-3p | MIMAT0040368 |
Rabbit | ocu-miR-656-3p | MIMAT0048550 |
Rhesus monkey | mml-miR-656-3p | MIMAT0006497 |
Tupaia chinensis | tch-miR-656 | MIMAT0036556 |
Stem-loop Details
Human
Bovine
Dasypus novemcinctus
Goat
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Tupaia chinensis