Assay Details
Gene Family ID: | MIPF0001805, mir-6505 |
Mature miRNA Sequence: | UUGGAAUAGGGGAUAUCUCAGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6505-5p | MIMAT0025466 |
Rhesus monkey | mml-miR-6505-5p | MIMAT0028248 |
Stem-loop Details
Human
Rhesus monkey