Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGGUUGCCUGUGAGGUGUUCA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-656-5p | MIMAT0026627 |
Dasypus novemcinctus | dno-miR-656-5p | MIMAT0047992 |
Guinea pig | cpo-miR-656-5p | MIMAT0047363 |
Pteropus alecto | pal-miR-656-5p | MIMAT0040367 |
Rabbit | ocu-miR-656-5p | MIMAT0048549 |
Stem-loop Details
Human
Dasypus novemcinctus
Guinea pig
Pteropus alecto
Rabbit