Assay Details
Mature miRNA Sequence: | UAGAGAGGGGAAGGAUGUGAUGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6758-5p | MIMAT0027416 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6758-5p | ||
---|---|---|---|
miRBase Accession Number: | MI0022603 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.12: 57512688 - 57512750 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UAGAGAGGGGAAGGAUGUGAUGU | ||
Species: | Human | ||
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UAGAGAGGGGAAGGAUGUGAUGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6758-5p | MIMAT0027416 |
Stem-loop ID | hsa-mir-6758 |
Stem-loop Accession # | MI0022603 |
Stem-loop Sequence | UGGGCUAGAGAGGGGAAGGAUGUGAUGUGAGCAGAUGGUUCUCACUCAUUCUCCUCUGUCCAG |
Chromosome Location | Chr. 12 - 57512688 - 57512750 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027416
miRBase ID: hsa-miR-6758-5p Mature miRNA Sequence: UAGAGAGGGGAAGGAUGUGAUGU Chromosome Location: Chr. 12 - 57512688 - 57512750 [+] on Build GRCh38 |