Assay Details
Mature miRNA Sequence: | CCUGAGACCCUAGUUCCAC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4329 | MIMAT0016923 |
Stem-loop Details
Human
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4329 | ||
---|---|---|---|
miRBase Accession Number: | MI0015901 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.X: 112780718 - 112780788 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | CCUGAGACCCUAGUUCCAC | ||
Species: | Human | ||
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | CCUGAGACCCUAGUUCCAC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4329 | MIMAT0016923 |
Stem-loop ID | hsa-mir-4329 |
Stem-loop Accession # | MI0015901 |
Stem-loop Sequence | UAGAGAGGAAGGUGUACCAGGGUUUUGGAGUUUUUUUUUCCUCCUGAGACCCUAGUUCCACAUUCUGGAGC |
Chromosome Location | Chr. X - 112780718 - 112780788 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0016923
miRBase ID: hsa-miR-4329 Mature miRNA Sequence: CCUGAGACCCUAGUUCCAC Chromosome Location: Chr. X - 112780718 - 112780788 [-] on Build GRCh38 |