Assay Details
Gene Family ID: | MIPF0000547, mir-649 |
Mature miRNA Sequence: | AAACCUGUGUUGUUCAAGAGUC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-649 | MIMAT0003319 |
Pan troglodytes | ptr-miR-649 | MIMAT0008300 |
Rhesus monkey | mml-miR-649 | MIMAT0006487 |
Stem-loop Details
Human
Pan troglodytes
Rhesus monkey