Assay Details
Target Gene Details
Entrez Gene ID: | 76485 |
Gene Name: | glycosyltransferase 8 domain containing 1 |
Gene Aliases: |
2410004H05Rik, 5430414N14Rik, AI450005 |
Location: |
Chr.14:31001347-31012441 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glt8d1 | NM_001165930.1 | NP_001159402.1 | ||
NM_029626.2 | NP_083902.2 |
Target Gene Details
Entrez Gene ID: | 30877 |
Gene Name: | guanine nucleotide binding protein-like 3 (nucleolar) |
Gene Aliases: |
Ns |
Location: |
Chr.14:31012433-31019131 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnl3 | NM_153547.6 | NP_705775.2 | ||
NR_104298.1 | ||||
AK077523.1 | BAC36844.1 | |||
AK167183.1 | BAE39318.1 | |||
AY181025.1 | AAO19472.1 | |||
AY185498.1 | AAO19473.1 | |||
BC037996.1 | AAH37996.2 |
More Information
Set Membership: |
Intragenic Non-exonic Validated |