Assay Details
Target Gene Details
Entrez Gene ID: | 19157 |
Gene Name: | cytohesin 1 |
Gene Aliases: |
CLM1, CTH-1, CYTIP, Pscd1, mKIAA4240 |
Location: |
Chr.11:118164166-118248592 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyth1 | NM_001112699.1 | NP_001106169.1 | ||
NM_001112700.1 | NP_001106170.1 | |||
NM_011180.3 | NP_035310.2 | |||
AB013464.1 | BAA33428.1 | |||
AB013465.1 | ||||
AB035538.1 | BAB13509.1 | |||
AK135303.1 | ||||
AK158216.1 | BAE34408.1 | |||
AK172296.1 | BAE42931.1 | |||
AK220302.1 | BAD90227.1 | |||
AY126516.1 | AAM95454.1 | |||
BC057974.1 | AAH57974.1 | |||
BY204859.1 | ||||
CJ131185.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic Validated |