Assay Details
Target Gene Details
Entrez Gene ID: | 27397 |
Gene Name: | mitochondrial ribosomal protein L17 |
Gene Aliases: |
MRP-L26, Rpml26 |
Location: |
Chr.7:105803782-105811087 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mrpl17 | NM_025301.2 | NP_079577.1 | ||
AK007438.1 | ||||
AK007828.1 | BAB25288.1 | |||
AK012906.1 | ||||
AK088838.1 | ||||
AK144092.1 | BAE25695.1 | |||
AK147957.1 | BAE28251.1 | |||
BC010718.1 | AAH10718.1 | |||
BY098205.1 |
More Information
Set Membership: |
Intragenic Non-exonic Validated |