Assay Details
Target Gene Details
Entrez Gene ID: | 59002 |
Gene Name: | WD repeat containing, antisense to Trp73 |
Gene Aliases: |
2610044M17Rik, 5330425N03Rik, Dd57, Wdr8 |
Location: |
Chr.4:154142357-154168259 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wrap73 | NM_021499.2 | NP_067474.2 | ||
AB034911.2 | BAA92311.1 | |||
AK011773.1 | ||||
AK051688.1 | BAC34722.1 | |||
AK156969.1 | BAE33920.1 | |||
AK160331.1 | BAE35741.1 | |||
AK172690.1 | BAE43131.1 |