Assay Details
Target Gene Details
Entrez Gene ID: | 11806 |
Gene Name: | apolipoprotein A-I |
Gene Aliases: |
Alp-1, Apoa-1, Brp-14, Ltw-1, Lvtw-1, Sep-1, Sep-2, Sep2, apo-AI, apoA-I |
Location: |
Chr.9:46228630-46230469 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apoa1 | NM_009692.4 | 4 | 858 | NP_033822.2 |
AK076187.1 | 4 | 781 | BAC36241.1 | |
AK131864.1 | 4 | 780 | BAE20837.1 | |
AK149576.1 | 4 | 780 | BAE28968.1 | |
AK161536.1 | 4 | 780 | BAE36448.1 | |
BC012253.1 | 3 | 762 | AAH12253.1 | |
BC019837.1 | 4 | 780 | AAH19837.1 | |
BC091745.1 | 4 | 699 | AAH91745.1 | |
BU755007.1 | 1 | 144 | ||
L04151.1 | 3 | 771 | ||
X64262.1 | 4 | 780 | CAA45560.1 |
More Information
Set Membership: |
Intragenic Exonic |