Assay Details
Target Gene Details
Entrez Gene ID: | 394430 |
Gene Name: | UDP glycosyltransferase 1 family, polypeptide A10 |
Gene Aliases: |
A13, Ugt1a13 |
Location: |
Chr.1:88055411-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ugt1a10 | NM_201641.2 | 1 | 44 | NP_964003.2 |
AY227201.1 | AAP48600.1 | |||
BC089569.1 | 1 | 44 | AAH89569.1 |
More Information
Set Membership: |
Intragenic Exonic |