Assay Details
Target Gene Details
Entrez Gene ID: | 12752 |
Gene Name: | ceroid lipofuscinosis, neuronal 3, juvenile (Batten, Spielmeyer-Vogt disease) |
Gene Aliases: |
AI323623 |
Location: |
Chr.7:126571400-126585817 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cln3 | NM_001146311.1 | 17 | 1557 | NP_001139783.1 |
NM_009907.3 | 15 | 1445 | NP_034037.3 | |
AK078976.1 | 15 | 1429 | BAC37491.1 | |
AK134036.1 | 18 | 2080 | ||
BC058753.1 | 16 | 1381 | AAH58753.1 | |
BC060306.1 | 14 | 1327 | AAH60306.1 | |
BC080759.1 | 16 | 1339 | AAH80759.1 | |
U47106.1 | 15 | 1395 | AAC52957.1 | |
U68064.1 | 15 | 1415 | AAB07595.1 | |
U92811.1 | 15 | 1364 | AAB69983.1 |
More Information
Set Membership: |
Intragenic Exonic |