Assay Details
Target Gene Details
Entrez Gene ID: | 13884 |
Gene Name: | carboxylesterase 1C |
Gene Aliases: |
Ces-N, Ee-1, Ee1, Es-4, Es-N, Es1, Es4, EsN, PESN |
Location: |
Chr.8:93099015-93131283 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ces1c | NM_007954.4 | NP_031980.2 | ||
AI116924.1 | ||||
AI527022.1 | ||||
AK050444.1 | ||||
BC028907.1 | AAH28907.1 | |||
M57960.1 | AAA63297.1 |
More Information
Set Membership: |
Intragenic Non-exonic |