Assay Details
Target Gene Details
Entrez Gene ID: | 76633 |
Gene Name: | RIKEN cDNA 1700112E06 gene |
Gene Aliases: |
2700009F18Rik |
Location: |
Chr.14:22019503-23056088 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700112E06Rik | NM_028275.1 | NP_082551.1 | ||
AK007175.1 | BAB24884.1 | |||
AK153491.1 | BAE32039.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |