Assay Details
Target Gene Details
Entrez Gene ID: | 67808 |
Gene Name: | transformation related protein 63 regulated like |
Gene Aliases: |
1200015A19Rik, Svap30, Tprg1l, mover |
Location: |
Chr.4:154157485-154160712 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tprgl | NM_026388.2 | 1 | 193 | NP_080664.1 |
AK004777.1 | 1 | 176 | BAB23555.1 | |
AK155413.1 | 1 | 188 | BAE33252.1 | |
BC019544.1 | 1 | 157 | AAH19544.1 | |
BY739996.1 | 1 | 193 |
Target Gene Details
Entrez Gene ID: | 59002 |
Gene Name: | WD repeat containing, antisense to Trp73 |
Gene Aliases: |
2610044M17Rik, 5330425N03Rik, Dd57, Wdr8 |
Location: |
Chr.4:154142357-154168259 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wrap73 | AK160273.1 | |||
AK172690.1 | BAE43131.1 |