Assay Details
Target Gene Details
Entrez Gene ID: | 68197 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2 |
Gene Aliases: |
1810004I06Rik, 2010300P09Rik, G1, R75094 |
Location: |
Chr.7:97400003-97407800 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufc2 | NM_024220.2 | NP_077182.1 | ||
AI413456.1 | ||||
AK002373.1 | BAB22050.1 | |||
AK003380.2 | ||||
AK008502.1 | BAB25703.1 | |||
AK008889.1 | BAB25955.1 | |||
AK019002.1 | BAB31504.1 | |||
AK153547.1 | BAE32084.1 | |||
AK166594.1 | BAE38880.1 | |||
AK168342.1 | BAE40279.1 | |||
BC002097.1 | AAH02097.1 | |||
CK128102.1 |
More Information
Set Membership: |
Intragenic Non-exonic |