Assay Details
Target Gene Details
Entrez Gene ID: | 12028 |
Gene Name: | BCL2-associated X protein |
Gene Aliases: |
- |
Location: |
Chr.7:45461695-45466903 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bax | NM_007527.3 | 1 | 105 | NP_031553.1 |
AK013298.1 | BAB28776.1 | |||
AK149994.1 | BAE29222.1 | |||
AK159233.1 | 1 | 70 | BAE34919.1 | |
AY095934.1 | AAM34436.1 | |||
BC018228.1 | AAH18228.1 | |||
BC053380.1 | AAH53380.1 | |||
BY308683.1 | 1 | 85 | ||
CJ078904.1 | 1 | 106 | ||
L22472.1 | AAA03622.1 |