Assay Details
Target Gene Details
Entrez Gene ID: | 227485 |
Gene Name: | cadherin 19, type 2 |
Gene Aliases: |
- |
Location: |
Chr.1:110888002-110977593 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh19 | NM_001081386.1 | 5 | 922 | NP_001074855.1 |
AK148396.1 | 5 | 910 | BAE28530.1 | |
BC157927.1 | 5 | 913 |
More Information
Set Membership: |
Intragenic Exonic |