Assay Details
Target Gene Details
Entrez Gene ID: | 15184 |
Gene Name: | histone deacetylase 5 |
Gene Aliases: |
AI426555, Hdac4, mHDA1, mKIAA0600 |
Location: |
Chr.11:102195747-102230186 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac5 | NM_001077696.1 | 13 | 2162 | NP_001071164.1 |
NM_001284248.1 | 15 | 2674 | NP_001271177.1 | |
NM_001284249.1 | 14 | 2271 | NP_001271178.1 | |
NM_001284250.1 | 13 | 2159 | NP_001271179.1 | |
NM_010412.3 | 13 | 2159 | NP_034542.3 | |
AF006602.2 | 12 | 1872 | AAD09834.2 | |
AF207748.1 | 12 | 1791 | AAF31418.1 | |
AK122325.2 | 8 | 1198 | BAC65607.2 | |
AK139202.1 | 14 | 2245 | ||
AK145972.1 | 13 | 2125 | ||
AK146483.1 | 13 | 2098 | BAE27204.1 | |
AK155335.1 | 13 | 2133 | BAE33201.1 | |
AK156008.1 | 7 | 2996 | ||
AK157159.1 | 7 | 2996 | ||
AK182309.1 | 1 | 102 | ||
AK182416.1 | 1 | 102 | ||
AK191157.1 | 1 | 102 | ||
AK197372.1 | 1 | 131 | ||
AK198950.1 | 1 | 100 | ||
AK213216.1 | 1 | 102 | ||
BC060609.1 | 12 | 1953 | AAH60609.1 |
More Information
Set Membership: |
Intragenic Exonic |