Assay Details
Target Gene Details
Entrez Gene ID: | 66979 |
Gene Name: | polymerase (DNA-directed), epsilon 4 (p12 subunit) |
Gene Aliases: |
2400007P05Rik, 5830430F06Rik |
Location: |
Chr.6:82618925-82652936 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pole4 | NM_025882.3 | 4 | 718 | NP_080158.1 |
AK010135.1 | 4 | 713 | BAB26722.1 | |
AK010300.1 | 4 | 719 | BAB26833.1 | |
AK011595.1 | BAB27723.1 | |||
AK017956.1 | 4 | 702 | ||
AK145365.1 | ||||
AK160582.1 | 4 | 730 | BAE35889.1 | |
AK167631.1 | 4 | 708 | BAE39682.1 | |
AK207330.1 | ||||
AK207634.1 | 1 | 192 | ||
BC023189.1 | 4 | 696 | AAH23189.1 |
More Information
Set Membership: |
Intragenic Exonic |