Assay Details
Target Gene Details
Entrez Gene ID: | 17527 |
Gene Name: | MpV17 mitochondrial inner membrane protein |
Gene Aliases: |
Tg.Mpv17 |
Location: |
Chr.5:31140663-31154532 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpv17 | NM_001294322.1 | 7 | 547 | NP_001281251.1 |
NM_001294324.1 | 8 | 609 | NP_001281253.1 | |
NM_008622.6 | 8 | 580 | NP_032648.1 | |
BC013452.1 | 7 | 533 | AAH13452.1 | |
CF731561.1 | ||||
CK622051.1 | ||||
DV062332.1 | 8 | 662 | ||
DV660230.1 | 5 | 301 | ||
M36411.1 | 8 | 562 | AAA39736.1 |
More Information
Set Membership: |
Intragenic Exonic |