Assay Details
Target Gene Details
Entrez Gene ID: | 18519 |
Gene Name: | K(lysine) acetyltransferase 2B |
Gene Aliases: |
A930006P13Rik, AI461839, AW536563, Pcaf, p/CAF |
Location: |
Chr.17:53566853-53672721 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat2b | NM_001190846.1 | 18 | 2305 | NP_001177775.1 |
NM_020005.4 | 18 | 2859 | NP_064389.2 | |
AK030070.1 | 18 | 2306 | ||
AK132486.1 | 18 | 2560 | ||
AK156290.1 | 18 | 2859 | BAE33658.1 | |
BC082581.1 | 18 | 2515 | AAH82581.1 | |
BC138195.1 | 18 | 2527 | ||
BC145896.1 | 18 | 2527 |